Search
Search

NAF1| Gene Symbol: NAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128931.1 | NP_001122403.1 | 5 | 1012 | ||
| NM_138386.2 | NP_612395.2 | 5 | 1012 | |||
| XM_011532410.2 | XP_011530712.1 | 5 | 4479 | |||
| XM_011532411.2 | XP_011530713.1 | 6 | 1086 | |||
| GenBank mRNA | AK097352.1 | - | 5 | 922 | ||
| AK308601.1 | - | 5 | 981 | |||
| BC008207.1 | - | 5 | 1012 | 3447276 | ||