Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271006.1 | NP_001257935.1 | 5 | 393 | ||
NM_001271007.1 | NP_001257936.1 | 4 | 324 | |||
NM_144998.3 | NP_659435.2 | 4 | 339 | |||
XM_005256339.2 | XP_005256396.1 | 5 | 545 | |||
XM_017024326.1 | XP_016879815.1 | 3 | 746 | |||
XM_017024327.1 | XP_016879816.1 | 4 | 618 | |||
XM_017024328.1 | XP_016879817.1 | 4 | 487 | |||
XM_017024329.1 | XP_016879818.1 | 3 | 433 | |||
GenBank mRNA | BC009571.1 | - | 4 | 308 | 4333630 | |
BC011610.1 | - | 4 | 275 | |||
BI837753.1 | - | 4 | 307 | |||
BQ648002.1 | - | 1 | 28 | |||
CB110145.1 | - | 5 | 402 | |||
HY110543.1 | - | 5 | 383 | |||
U95006.1 | - | 4 | 293 | |||
U95007.1 | - | 4 | 278 |