Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAS6 SIRNA Assays ›
          Gene Symbol:
          GAS6
          Gene Name:
          growth arrest specific 6
          Gene Aliases:
          AXLLG, AXSF
          Chromosome Location:
          Chr.13: 113820549 - 113864103 on Build GRCh38
          UniGene ID:
          Hs.646346
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3929

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAS6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000820.3 NP_000811.1 6, 7 769
          GenBank mRNA AK126533.1 - 5, 6 2053
          AK290803.1 - 6, 7 658
          AK303145.1 - 3, 4 315
          BC038984.1 - 6, 7 717 5761791
          BX380508.2 - 6, 7 767
          L13720.1 - 6, 7 720

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          positive regulation of protein phosphorylation
          positive regulation of cytokine-mediated signaling pathway
          platelet degranulation
          positive regulation of glomerular filtration
          signal peptide processing
          protein phosphorylation
          ER to Golgi vesicle-mediated transport
          phagocytosis
          apoptotic process
          cell cycle arrest
          cell adhesion
          signal transduction
          enzyme linked receptor protein signaling pathway
          cell proliferation
          cellular response to starvation
          positive regulation of gene expression
          negative regulation of tumor necrosis factor-mediated signaling pathway
          macrophage cytokine production
          cell migration
          peptidyl-serine phosphorylation
          fusion of virus membrane with host plasma membrane
          viral genome replication
          platelet activation
          organ regeneration
          cell-substrate adhesion
          positive regulation of TOR signaling
          activation of protein kinase B activity
          negative regulation of interferon-gamma production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of protein import into nucleus, translocation
          cellular response to interferon-alpha
          cellular response to drug
          B cell chemotaxis
          regulation of growth
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          apoptotic cell clearance
          negative regulation of sequence-specific DNA binding transcription factor activity
          protein kinase B signaling
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          viral entry into host cell
          receptor-mediated virion attachment to host cell
          positive regulation of protein export from nucleus
          positive regulation of fibroblast proliferation
          negative regulation of interleukin-1 secretion
          positive regulation of phagocytosis
          leukocyte migration
          positive regulation of protein kinase B signaling
          positive regulation of protein tyrosine kinase activity
          negative regulation of biomineral tissue development
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          calcium ion transmembrane transport
          cellular response to vitamin K
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          protein targeting to plasma membrane
          extracellular matrix assembly
          dendritic cell differentiation
          hematopoietic stem cell migration to bone marrow
          negative regulation of oligodendrocyte apoptotic process
          negative regulation of interleukin-6 secretion
          negative regulation of fibroblast apoptotic process
          negative regulation of endothelial cell apoptotic process
          positive regulation of dendritic cell chemotaxis
          negative regulation of renal albumin absorption
          negative regulation of dendritic cell apoptotic process
          phosphatidylserine binding
          receptor binding
          voltage-gated calcium channel activity
          calcium ion binding
          protein binding
          protein tyrosine kinase activator activity
          receptor tyrosine kinase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          receptor agonist activity
          binding, bridging

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0