Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADD1 SIRNA Assays ›
          Gene Symbol:
          ADD1
          Gene Name:
          adducin 1
          Gene Aliases:
          ADDA
          Chromosome Location:
          Chr.4: 2843732 - 2930076 on Build GRCh38
          UniGene ID:
          Hs.183706
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 42990

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001119.4 NP_001110.2 2 264
          NM_001286645.1 NP_001273574.1 3 939
          NM_014189.3 NP_054908.2 2 264
          NM_014190.3 NP_054909.2 2 264
          NM_176801.2 NP_789771.1 2 264
          XM_005247933.2 XP_005247990.1 2 389
          XM_005247934.2 XP_005247991.1 2 353
          XM_005247935.2 XP_005247992.1 2 389
          XM_005247936.2 XP_005247993.1 2 389
          XM_005247937.2 XP_005247994.1 2 389
          XM_017007703.1 XP_016863192.1 2 114
          XM_017007704.1 XP_016863193.1 2 353
          XM_017007705.1 XP_016863194.1 2 353
          XM_017007706.1 XP_016863195.1 2 353
          XM_017007707.1 XP_016863196.1 1 111
          XM_017007708.1 XP_016863197.1 2 389
          XM_017007709.1 XP_016863198.1 2 353
          XM_017007710.1 XP_016863199.1 1 111
          GenBank mRNA AK295457.1 - 2 351
          AK309186.1 - 2 142
          BC013393.2 - 2 199 3532276
          BC042998.1 - 3 944 5297337
          BI460192.1 - 2 287
          CR995783.1 - 2 239
          D44632.1 - 2 281
          L07261.1 - 1 78

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          in utero embryonic development
          cell volume homeostasis
          hemoglobin metabolic process
          actin cytoskeleton organization
          erythrocyte differentiation
          positive regulation of protein binding
          multicellular organism growth
          IRE1-mediated unfolded protein response
          homeostasis of number of cells within a tissue
          barbed-end actin filament capping
          actin filament bundle assembly
          transmembrane transport
          cell-cell adhesion
          actin binding
          structural molecule activity
          calmodulin binding
          transcription factor binding
          spectrin binding
          protein homodimerization activity
          poly(A) RNA binding
          protein heterodimerization activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0