Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCOR SIRNA Assays ›
          Gene Symbol:
          BCOR
          Gene Name:
          BCL6 corepressor
          Gene Aliases:
          ANOP2, MAA2, MCOPS2
          Chromosome Location:
          Chr.X: 40051246 - 40177390 on Build GRCh38
          UniGene ID:
          Hs.659681
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44447

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCOR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123383.1 NP_001116855.1 2, 3 457
          NM_001123384.1 NP_001116856.1 2, 3 457
          NM_001123385.1 NP_001116857.1 2, 3 386
          NM_017745.5 NP_060215.4 2, 3 386
          XM_005272616.1 XP_005272673.1 2, 3 518
          XM_005272618.3 XP_005272675.1 2, 3 184
          XM_005272619.3 XP_005272676.1 2, 3 844
          XM_005272620.3 XP_005272677.1 2, 3 844
          XM_006724536.3 XP_006724599.1 2, 3 1620
          XM_011543929.2 XP_011542231.1 2, 3 294
          XM_011543930.1 XP_011542232.1 2, 3 307
          XM_011543931.2 XP_011542233.1 2, 3 417
          XM_017029615.1 XP_016885104.1 2, 3 1620
          XM_017029616.1 XP_016885105.1 2, 3 844
          GenBank mRNA AB046795.2 - 1, 2 112
          AF317391.1 - 2, 3 345
          AF317392.1 - 2, 3 345
          AK056538.1 - 2, 3 211
          AL698729.1 - 2, 3 386
          AY316592.1 - 2, 3 345
          BC114220.1 - 2, 3 457 4384724
          BC128456.1 - 1, 2 122

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of histone H3-K36 methylation
          transcription, DNA-templated
          heart development
          negative regulation of bone mineralization
          histone H2A monoubiquitination
          odontogenesis
          negative regulation of transcription, DNA-templated
          negative regulation of histone H3-K4 methylation
          palate development
          specification of axis polarity
          negative regulation of tooth mineralization
          transcription corepressor activity
          ubiquitin-protein transferase activity
          protein binding
          transcription factor binding
          heat shock protein binding
          histone deacetylase binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0