Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASK SIRNA Assays ›
          Gene Symbol:
          CASK
          Gene Name:
          calcium/calmodulin dependent serine protein kinase
          Gene Aliases:
          CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
          Chromosome Location:
          Chr.X: 41514934 - 41923525 on Build GRCh38
          UniGene ID:
          Hs.495984
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 447
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126054.2 NP_001119526.1 13, 14 1287
          NM_001126055.2 NP_001119527.1 12, 13 1269
          NM_003688.3 NP_003679.2 13, 14 1287
          XM_005272686.4 XP_005272743.1 12, 13 1760
          XM_006724566.3 XP_006724629.1 12, 13 1760
          XM_011543993.2 XP_011542295.1 12, 13 1339
          XM_011543994.2 XP_011542296.1 12, 13 1339
          XM_011543995.2 XP_011542297.1 12, 13 1339
          XM_011543996.2 XP_011542298.1 12, 13 1339
          XM_011543997.2 XP_011542299.1 7, 8 998
          GenBank mRNA AB039327.2 - 1, 2 445
          AB208859.1 - 13, 14 1231
          AF032119.1 - 13, 14 1268
          AF035582.1 - 12, 13 1238
          AF262404.1 - 8, 9 727
          AF262405.1 - 11, 12 1145
          AK313399.1 - 12, 13 1422
          AY705392.1 - 13, 14 1247
          BC117311.1 - 13, 14 1260 40125862
          BC143454.1 - 13, 14 1260 9051962
          BC143456.1 - 12, 13 1242 9051964

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_022184, U47110.
          This siRNA also targets the following transcripts in Mouse: NM_001284505, NM_009806, NM_001284504, NM_001284503, BC141292, and others.

          Matching Taqman Gene Expression Assay:


          Hs00177620_m1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell-matrix adhesion
          protein phosphorylation
          cell adhesion
          neurotransmitter secretion
          negative regulation of keratinocyte proliferation
          positive regulation of transcription from RNA polymerase II promoter
          GMP metabolic process
          GDP metabolic process
          negative regulation of wound healing
          calcium ion import
          positive regulation of calcium ion import
          negative regulation of cellular response to growth factor stimulus
          guanylate kinase activity
          protein serine/threonine kinase activity
          protein binding
          calmodulin binding
          ATP binding
          neurexin family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0