Search
Search

FRMD8| Gene Symbol: FRMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300832.1 | NP_001287761.1 | 6, 7 | 821 | ||
| NM_001300833.1 | NP_001287762.1 | 6, 7 | 887 | |||
| NM_031904.3 | NP_114110.1 | 7, 8 | 989 | |||
| GenBank mRNA | AF334946.1 | - | 8, 9 | 923 | ||
| AK074850.1 | - | 6, 7 | 754 | |||
| AK291678.1 | - | 7, 8 | 902 | |||
| AK304363.1 | - | 6, 7 | 800 | |||
| BC044658.1 | - | 7, 8 | 907 | 5759383 | ||
| BC051695.1 | - | 7, 8 | 902 | 5298820 | ||