Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP2B2 SIRNA Assays ›
Gene Symbol:
Atp2b2
Gene Name:
ATPase, Ca++ transporting, plasma membrane 2
Gene Aliases:
PMCA2
Chromosome Location:
Chr.4: 145704779 - 145948997 on Build Rnor_6.0
UniGene ID:
Rn.90982
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
47868
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Atp2b2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012508.5
NP_036640.1
2694
GenBank mRNA
J03754.1
-
2700
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001036684, NM_009723, AF053471, AK147240, AK147369, and others.
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
neural retina development
calcium ion transport
cellular calcium ion homeostasis
organelle organization
brain development
lactation
sensory perception of sound
locomotory behavior
metabolic process
regulation of cell size
cerebellum development
cerebellar Purkinje cell layer morphogenesis
cerebellar Purkinje cell differentiation
cerebellar granule cell differentiation
neuron differentiation
locomotion
serotonin metabolic process
inner ear morphogenesis
otolith mineralization
cGMP metabolic process
regulation of synaptic plasticity
inner ear development
synapse organization
neuromuscular process controlling balance
detection of mechanical stimulus involved in sensory perception of sound
regulation of cytosolic calcium ion concentration
positive regulation of calcium ion transport
auditory receptor cell stereocilium organization
inner ear receptor cell differentiation
calcium ion transmembrane transport
cochlea development
calcium ion export
calcium-transporting ATPase activity
calcium ion binding
calmodulin binding
ATP binding
protein C-terminus binding
PDZ domain binding
calcium-dependent ATPase activity
acetylcholine receptor binding
glutamate receptor binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0