Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRP1 SIRNA Assays ›
          Gene Symbol:
          NRP1
          Gene Name:
          neuropilin 1
          Gene Aliases:
          BDCA4, CD304, NP1, NRP, VEGF165R
          Chromosome Location:
          Chr.10: 33177491 - 33334905 on Build GRCh38
          UniGene ID:
          Hs.131704
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4820

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024628.2 NP_001019799.1 3 877
          NM_001024629.2 NP_001019800.1 3 877
          NM_001244972.1 NP_001231901.1 3 877
          NM_001244973.1 NP_001231902.1 3 877
          NM_003873.5 NP_003864.4 3 877
          XM_006717521.1 XP_006717584.1 3 651
          XM_006717522.1 XP_006717585.1 3 651
          XM_006717523.2 XP_006717586.1 3 651
          XM_006717524.1 XP_006717587.1 3 651
          XM_006717525.1 XP_006717588.1 3 651
          XM_006717526.1 XP_006717589.1 3 651
          XM_011519755.2 XP_011518057.1 3 651
          XM_011519756.1 XP_011518058.1 3 651
          XM_017016865.1 XP_016872354.1 3 651
          GenBank mRNA AB209641.1 - 3 505
          AF016050.1 - 3 354
          AF018956.1 - 3 354
          AF145712.1 - 3 354
          AF268691.1 - 3 494
          AF280547.1 - 3 354
          AK292822.1 - 3 543
          AY249243.1 - 3 354
          BC007533.1 - 3 614 2958475
          BC007737.2 - 3 494 4122289
          BT006995.1 - 3 354
          BX510902.1 - 3 541
          BX648025.1 - 4 901
          BX648250.1 - 3 846
          CR749333.1 - 3 878

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          patterning of blood vessels
          neuron migration
          positive regulation of endothelial cell proliferation
          sprouting angiogenesis
          cell migration involved in sprouting angiogenesis
          signal transduction
          cell-cell signaling
          axon guidance
          axonal fasciculation
          response to wounding
          organ morphogenesis
          positive regulation of endothelial cell migration
          positive regulation of smooth muscle cell migration
          dendrite development
          facial nerve structural organization
          trigeminal nerve structural organization
          vestibulocochlear nerve structural organization
          nerve development
          branchiomotor neuron axon guidance
          gonadotrophin-releasing hormone neuronal migration to the hypothalamus
          retinal ganglion cell axon guidance
          cellular response to hepatocyte growth factor stimulus
          endothelial cell chemotaxis
          cellular response to vascular endothelial growth factor stimulus
          ventral trunk neural crest cell migration
          VEGF-activated neuropilin signaling pathway
          otic placode formation
          negative regulation of neuron apoptotic process
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          hepatocyte growth factor receptor signaling pathway
          positive regulation of axon extension involved in axon guidance
          negative regulation of axon extension involved in axon guidance
          artery morphogenesis
          axon extension involved in axon guidance
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          positive regulation of cytokine activity
          axonogenesis involved in innervation
          regulation of vesicle-mediated transport
          dichotomous subdivision of terminal units involved in salivary gland branching
          angiogenesis involved in coronary vascular morphogenesis
          coronary artery morphogenesis
          retina vasculature morphogenesis in camera-type eye
          renal artery morphogenesis
          sympathetic ganglion development
          trigeminal ganglion development
          positive regulation of ERK1 and ERK2 cascade
          semaphorin-plexin signaling pathway
          commissural neuron axon guidance
          regulation of retinal ganglion cell axon guidance
          endothelial tip cell fate specification
          sensory neuron axon guidance
          sympathetic neuron projection extension
          sympathetic neuron projection guidance
          neural crest cell migration involved in autonomic nervous system development
          toxin transport
          semaphorin-plexin signaling pathway involved in neuron projection guidance
          semaphorin-plexin signaling pathway involved in axon guidance
          positive regulation of retinal ganglion cell axon guidance
          VEGF-activated neuropilin signaling pathway involved in axon guidance
          protein localization to early endosome
          facioacoustic ganglion development
          dorsal root ganglion morphogenesis
          negative regulation of extrinsic apoptotic signaling pathway
          vascular endothelial growth factor-activated receptor activity
          protein binding
          heparin binding
          coreceptor activity
          semaphorin receptor activity
          growth factor binding
          cytokine binding
          vascular endothelial growth factor binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0