Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPM1 SIRNA Assays ›
          Gene Symbol:
          Npm1
          Gene Name:
          nucleophosmin (nucleolar phosphoprotein B23, numatrin)
          Gene Aliases:
          B23NP
          Chromosome Location:
          Chr.10: 18080949 - 18091063 on Build Rnor_6.0
          UniGene ID:
          Rn.54537
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 48790

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Npm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012992.4 NP_037124.1 2 173
          XM_008767546.1 XP_008765768.1 2 286
          GenBank mRNA BC060579.1 - 2 177 6888417
          BC088088.1 - 2 163 7382200
          J03969.1 - 2 126
          J04943.1 - 2 155

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit export from nucleus
          ribosomal small subunit export from nucleus
          DNA repair
          nucleosome assembly
          rRNA export from nucleus
          cell volume homeostasis
          nucleocytoplasmic transport
          centrosome cycle
          cell aging
          protein localization
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of centrosome duplication
          positive regulation of centrosome duplication
          negative regulation of centrosome duplication
          cell growth
          positive regulation of cellular biosynthetic process
          protein destabilization
          regulation of endodeoxyribonuclease activity
          ribosomal large subunit biogenesis
          ribosomal small subunit biogenesis
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          regulation of DNA damage response, signal transduction by p53 class mediator
          regulation of neuron apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of translation
          positive regulation of DNA replication
          positive regulation of protein kinase activity
          regulation of centriole replication
          negative regulation of mRNA splicing, via spliceosome
          positive regulation of DNA metabolic process
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          protein homooligomerization
          regulation of cell cycle
          regulation of endoribonuclease activity
          regulation of eIF2 alpha phosphorylation by dsRNA
          DNA binding
          transcription coactivator activity
          RNA binding
          protein kinase inhibitor activity
          phosphatidylinositol-3,4,5-trisphosphate binding
          rRNA binding
          enzyme binding
          protein kinase binding
          Tat protein binding
          histone binding
          protein homodimerization activity
          ribosomal large subunit binding
          ribosomal small subunit binding
          poly(A) RNA binding
          protein heterodimerization activity
          NF-kappaB binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0