Search
Search

CRADD| Gene Symbol: CRADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320099.1 | NP_001307028.1 | 3 | 1287 | ||
| NM_003805.4 | NP_003796.1 | 3 | 1077 | |||
| GenBank mRNA | BC017042.1 | - | 3 | 1049 | 3849848 | |
| BC037905.1 | - | 3 | 1291 | 5276510 | ||
| U84388.1 | - | 3 | 1008 | |||