Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019269.1 | NP_062142.1 | 9, 10 | 1685 | ||
GenBank mRNA | AB017260.1 | - | 9, 10 | 1685 | ||
AF110416.1 | - | 9, 10 | 1685 | |||
AJ001933.1 | - | 9, 10 | 1671 | |||
BC166421.1 | - | 8, 9 | 1434 | 9089728 |