Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030990.2 | NP_112252.1 | 3 | 448 | ||
XM_006257293.2 | XP_006257355.1 | 6 | 835 | |||
XM_008773401.1 | XP_008771623.1 | 6 | 834 | |||
GenBank mRNA | BC093596.1 | - | 2 | 309 | 7314758 | |
FQ211530.1 | - | 3 | 463 | |||
FQ212519.1 | - | 2 | 162 | |||
FQ212680.1 | - | 2 | 162 | |||
FQ213540.1 | - | 2 | 162 | |||
FQ213548.1 | - | 2 | 162 | |||
FQ213841.1 | - | 2 | 162 | |||
FQ213943.1 | - | 2 | 162 | |||
FQ214012.1 | - | 2 | 162 | |||
FQ214050.1 | - | 2 | 162 | |||
FQ214153.1 | - | 2 | 162 | |||
FQ214556.1 | - | 2 | 162 | |||
M11185.1 | - | 3 | 423 | |||
X02809.1 | - | 3 | 352 |