Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033235.1 | NP_150238.1 | 6 | 656 | ||
GenBank mRNA | AF093773.1 | - | 6 | 656 | ||
BC059124.1 | - | 6 | 697 | 6921518 | ||
FQ212702.1 | - | 6 | 700 | |||
FQ215129.1 | - | 6 | 711 | |||
FQ215603.1 | - | 6 | 695 | |||
FQ217819.1 | - | 700 | ||||
FQ224891.1 | - | 6 | 698 |