Search
Search

Adssl1| Gene Symbol: Adssl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007421.2 | NP_031447.1 | 2, 3 | 359 | ||
| GenBank mRNA | AK146907.1 | - | 2, 3 | 362 | ||
| AK149634.1 | - | 2, 3 | 333 | |||
| AK150874.1 | - | 2, 3 | 330 | |||
| AK196699.1 | - | 82 | ||||
| BC039943.1 | - | 2, 3 | 321 | 4014672 | ||
| M74495.1 | - | 2, 3 | 317 | |||