Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALOX12 SIRNA Assays ›
          Gene Symbol:
          Alox12
          Gene Name:
          arachidonate 12-lipoxygenase
          Gene Aliases:
          9930022G08Rik, Alox12p, P-12LO
          Chromosome Location:
          Chr.11: 70241455 - 70255386 on Build GRCm38
          UniGene ID:
          Mm.12286
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 60069

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Alox12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007440.4 NP_031466.2 1, 2 215
          GenBank mRNA AK036898.1 - 1, 2 213
          AK036960.1 - 2 171
          AK079232.1 - 1, 2 216
          AK087724.1 - 1, 2 216
          AK172216.1 - 1, 2 213
          AK172488.1 - 1, 2 216
          BC152329.1 - 1, 2 220 40141477
          BC152403.1 - 1, 2 220 40141482
          S80446.1 - 1, 2 227
          U04334.1 - 1, 2 146

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          movement of cell or subcellular component
          positive regulation of cell proliferation
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of cell death
          arachidonic acid metabolic process
          lipoxygenase pathway
          fatty acid oxidation
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of vasodilation
          positive regulation of smooth muscle cell proliferation
          hepoxilin biosynthetic process
          positive regulation of mitochondrial depolarization
          oxidation-reduction process
          establishment of skin barrier
          negative regulation of platelet aggregation
          lipoxin A4 biosynthetic process
          lipoxin B4 biosynthetic process
          arachidonate 12-lipoxygenase activity
          iron ion binding
          juvenile hormone epoxide hydrolase activity
          linoleate 13S-lipoxygenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          hydrolase activity
          epoxide hydrolase B activity
          phenanthrene-9,10-epoxide hydrolase (9R,10R-forming) activity
          epoxide hydrolase A activity
          phenanthrene-3,4-epoxide hydrolase activity
          phenanthrene-1,2-epoxide hydrolase activity
          phenanthrene-9,10-epoxide hydrolase (9S,10S-forming) activity
          phenanthrene-epoxide hydrolase activity
          phenanthrene-9,10-epoxide hydrolase activity
          pyrene-4,5-epoxide hydrolase activity
          metal ion binding
          dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0