Search
Search

Kcnn4| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163510.1 | NP_001156982.1 | 2, 3 | 428 | ||
| NM_008433.4 | NP_032459.3 | 2, 3 | 428 | |||
| GenBank mRNA | AF042487.1 | - | 2, 3 | 294 | ||
| AF072884.1 | - | 2, 3 | 254 | |||
| AK010943.1 | - | 2, 3 | 429 | |||
| AK085752.1 | - | 2, 3 | 238 | |||
| AK155910.1 | - | 2, 3 | 393 | |||
| BC010274.1 | - | 2, 3 | 377 | 3589411 | ||