Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other XCL1 SIRNA Assays ›
          Gene Symbol:
          Xcl1
          Gene Name:
          chemokine (C motif) ligand 1
          Gene Aliases:
          AI661682, ATAC, LTN, Lptn, SCM-1, SCM-1a, Scyc1
          Chromosome Location:
          Chr.1: 164931648 - 164935510 on Build GRCm38
          UniGene ID:
          Mm.190
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62883

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Xcl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008510.1 NP_032536.1 3 383
          GenBank mRNA AK079820.1 - 3 396
          BC062249.1 - 3 383 3974601
          D43769.1 - 3 383
          U15607.1 - 3 381

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of leukocyte chemotaxis
          negative regulation of T cell cytokine production
          positive regulation of T cell cytokine production
          positive regulation of immune response to tumor cell
          chemotaxis
          immune response
          cell-cell signaling
          response to virus
          positive regulation of T cell chemotaxis
          negative regulation of interferon-gamma production
          negative regulation of interleukin-2 production
          positive regulation of interleukin-10 production
          mature natural killer cell chemotaxis
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of innate immune response
          negative regulation of transcription, DNA-templated
          regulation of inflammatory response
          release of sequestered calcium ion into cytosol
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to interleukin-4
          cellular response to transforming growth factor beta stimulus
          positive regulation of transforming growth factor beta production
          positive regulation of granzyme B production
          positive regulation of neutrophil chemotaxis
          positive regulation of thymocyte migration
          positive regulation of natural killer cell chemotaxis
          positive regulation of granzyme A production
          negative regulation of T-helper 1 cell activation
          positive regulation of B cell chemotaxis
          positive regulation of T-helper 2 cell cytokine production
          positive regulation of T-helper 1 cell cytokine production
          positive regulation of immunoglobulin production in mucosal tissue
          negative regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of CD8-positive, alpha-beta T cell proliferation
          cytokine activity
          chemokine activity
          chemokine receptor binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0