Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010851.2 | NP_034981.1 | 1, 2 | 419 | ||
GenBank mRNA | AK090107.1 | - | 1, 2 | 382 | ||
AK150083.1 | - | 1, 2 | 302 | |||
AK151748.1 | - | 1, 2 | 375 | |||
AK152596.1 | - | 1, 2 | 404 | |||
AK167929.1 | - | 1, 2 | 399 | |||
AK186240.1 | - | 1, 2 | 187 | |||
BC005591.1 | - | 1, 2 | 419 | 3482246 | ||
BC051075.1 | - | 1, 2 | 366 | |||
BC058787.1 | - | 1, 2 | 354 | 6309465 | ||
DQ790885.1 | - | 1, 2 | 338 | |||
U84409.1 | - | 1, 2 | 400 | |||
X51397.1 | - | 1, 2 | 288 |