Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NF2 SIRNA Assays ›
          Gene Symbol:
          Nf2
          Gene Name:
          neurofibromatosis 2
          Gene Aliases:
          merlin
          Chromosome Location:
          Chr.11: 4757650 - 4849544 on Build GRCm38
          UniGene ID:
          Mm.297109
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68422

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nf2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252250.1 NP_001239179.1 5, 6 1082
          NM_001252251.1 NP_001239180.1 5, 6 1082
          NM_001252252.1 NP_001239181.1 5, 6 1082
          NM_001252253.1 NP_001239182.1 5, 6 547
          NM_010898.4 NP_035028.2 5, 6 1082
          GenBank mRNA AK017343.1 - 5, 6 548
          AK045998.1 - 5, 6 904
          AK167685.1 - 2, 3 173
          BC005442.1 - 5, 6 948 3496053
          DV654856.1 - 2, 3 140
          L22987.1 - 5, 6 540
          L22988.1 - 5, 6 540
          L22989.1 - 5, 6 540
          L27090.1 - 5, 6 533
          L27105.1 - 5, 6 533
          L28176.1 - 5, 6 840
          X74671.1 - 5, 6 1099

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013193, XM_006251191, XM_006251188, U61772.

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          negative regulation of protein kinase activity
          ectoderm development
          brain development
          negative regulation of DNA replication
          negative regulation of cell proliferation
          Schwann cell proliferation
          regulation of gliogenesis
          hippocampus development
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          negative regulation of cell growth
          regulation of protein stability
          regulation of hippo signaling
          regulation of cell proliferation
          odontogenesis of dentin-containing tooth
          negative regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of MAPK cascade
          cell-cell junction organization
          positive regulation of cell differentiation
          negative regulation of JAK-STAT cascade
          regulation of neurogenesis
          positive regulation of stress fiber assembly
          lens fiber cell differentiation
          regulation of stem cell proliferation
          regulation of protein localization to nucleus
          regulation of neural precursor cell proliferation
          actin binding
          integrin binding
          protein binding
          beta-catenin binding
          cytoskeletal protein binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0