Search
Search

Ctss| Gene Symbol: Ctss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267695.2 | NP_001254624.2 | 4 | 472 | ||
| NM_021281.3 | NP_067256.4 | 4 | 469 | |||
| GenBank mRNA | AF038546.1 | - | 4 | 411 | ||
| AJ002386.1 | - | 4 | 424 | |||
| AK028366.1 | - | 4 | 471 | |||
| AK150274.1 | - | 4 | 403 | |||
| AK150758.1 | - | 3 | 315 | |||
| AK150842.1 | - | 3 | 315 | |||
| AK152191.1 | - | 4 | 422 | |||
| AK152618.1 | - | 3 | 315 | |||
| AK153540.1 | - | 1 | 71 | |||
| BC002125.1 | - | 4 | 378 | 3495719 | ||
| BC011104.1 | - | 4 | 386 | |||