Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294269.1 | NP_001281198.1 | 2 | 697 | ||
NM_025396.3 | NP_079672.1 | 2 | 413 | |||
GenBank mRNA | AK003134.1 | - | 2 | 410 | ||
AK003985.1 | - | 2 | 414 | |||
AK028344.1 | - | 1 | 1971 | |||
AK036059.1 | - | 414 | ||||
AK172066.1 | - | 2 | 414 | |||
BC006594.1 | - | 2 | 387 | 3964951 | ||
BE654173.1 | - | 2 | 275 |