Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 12 | 1589 | ||
GenBank mRNA | AK004717.1 | - | 10 | 2551 | ||
AK084948.1 | - | 12 | 1589 | |||
AK085327.1 | - | 1 | 1620 | |||
AK153976.1 | - | 12 | 1560 | |||
AK154352.1 | - | 12 | 1531 | |||
AK167020.1 | - | 12 | 1538 | |||
AK172708.1 | - | 12 | 1557 | |||
AK188535.1 | - | 2 | 48 | |||
AK207344.1 | - | 4 | 296 | |||
BC048397.1 | - | 12 | 1568 | 6396815 |