Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SDR42E1 SIRNA Assays ›
Gene Symbol:
Sdr42e1
Gene Name:
short chain dehydrogenase/reductase family 42E, member 1
Gene Aliases:
4632417N05Rik, AI117603
Chromosome Location:
Chr.8: 117661399 - 117673718 on Build GRCm38
UniGene ID:
Mm.296302
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
83154
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sdr42e1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_028725.3
NP_083001.1
3
940
GenBank mRNA
AK014586.1
-
3
942
BC038819.1
-
3
679
3668862
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydratase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
biological_process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
molecular_function
3-beta-hydroxy-delta5-steroid dehydrogenase activity
aldo-keto reductase (NADP) activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
3-ketoglucose-reductase activity
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0