Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCSK9 SIRNA Assays ›
Gene Symbol:
Pcsk9
Gene Name:
proprotein convertase subtilisin/kexin type 9
Gene Aliases:
AI415265, AI747682, FH3, HCHOLA3, Narc1, PC9
Chromosome Location:
Chr.4: 106442334 - 106464325 on Build GRCm38
UniGene ID:
Mm.133268
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
93426
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pcsk9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_153565.2
NP_705793.1
12
2799
GenBank mRNA
AK043077.1
-
12
2650
AK149520.1
-
12
2578
BC038085.1
-
12
2814
4502199
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of receptor recycling
positive regulation of receptor internalization
proteolysis
lipid metabolic process
triglyceride metabolic process
phospholipid metabolic process
apoptotic process
lysosomal transport
steroid metabolic process
cholesterol metabolic process
cellular response to starvation
regulation of receptor activity
negative regulation of low-density lipoprotein particle clearance
protein processing
protein autoprocessing
neuron differentiation
low-density lipoprotein receptor particle metabolic process
low-density lipoprotein particle receptor catabolic process
regulation of low-density lipoprotein particle receptor catabolic process
positive regulation of low-density lipoprotein particle receptor catabolic process
cellular response to insulin stimulus
lipoprotein metabolic process
cholesterol homeostasis
regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
negative regulation of receptor activity
negative regulation of sodium ion transmembrane transporter activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity
sodium channel inhibitor activity
low-density lipoprotein particle binding
receptor inhibitor activity
apolipoprotein binding
very-low-density lipoprotein particle binding
apolipoprotein receptor binding
protein self-association
poly(A) RNA binding
low-density lipoprotein particle receptor binding
very-low-density lipoprotein particle receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ht48x:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0