Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYNC1I2 SIRNA Assays ›
Gene Symbol:
DYNC1I2
Gene Name:
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases:
DIC74, DNCI2, IC2
Chromosome Location:
Chr.2: 171687409 - 171750158 on Build GRCh38
UniGene ID:
Hs.546250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9926
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYNC1I2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271785.1
NP_001258714.1
8, 9
810
NM_001271786.1
NP_001258715.1
6, 7
803
NM_001271787.1
NP_001258716.1
6, 7
786
NM_001271788.1
NP_001258717.1
6, 7
749
NM_001271789.1
NP_001258718.1
6, 7
732
NM_001271790.1
NP_001258719.1
6, 7
749
NM_001320882.1
NP_001307811.1
7, 8
809
NM_001320883.1
NP_001307812.1
7, 8
792
NM_001320884.1
NP_001307813.1
6, 7
732
NM_001378.2
NP_001369.1
8, 9
827
XM_006712347.3
XP_006712410.1
7, 8
733
XM_017003525.1
XP_016859014.1
7, 8
768
GenBank mRNA
AB209014.1
-
4, 5
752
AF134477.1
-
6, 7
621
AF250307.1
-
6, 7
641
AK055491.1
-
8, 9
764
AK096579.1
-
8, 9
728
AK225759.1
-
6, 7
695
AK298561.1
-
6, 7
646
AK308008.1
-
6, 7
604
AK316119.1
-
6, 7
666
AL832832.1
-
2, 3
119
AY037160.1
-
6, 7
657
BC015038.2
-
6, 7
593
3921299
BC091498.1
-
6, 7
677
30386904
BC109375.1
-
6, 7
634
4608676
BT007130.1
-
5, 6
521
BX537412.1
-
6, 7
689
DQ786324.1
-
2, 3
144
U39575.1
-
4, 5
442
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
ER to Golgi vesicle-mediated transport
microtubule-based movement
viral process
antigen processing and presentation of exogenous peptide antigen via MHC class II
microtubule motor activity
protein binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0