Search
Search

KPNA5| Gene Symbol: KPNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002269.2 | NP_002260.2 | 3 | 345 | ||
| XM_011535805.1 | XP_011534107.1 | 4 | 523 | |||
| XM_017010838.1 | XP_016866327.1 | 4 | 523 | |||
| XM_017010839.1 | XP_016866328.1 | 3 | 326 | |||
| XM_017010840.1 | XP_016866329.1 | 4 | 483 | |||
| XM_017010842.1 | XP_016866331.1 | 3 | 326 | |||
| XM_017010843.1 | XP_016866332.1 | 4 | 450 | |||
| GenBank mRNA | AK314206.1 | - | 3 | 338 | ||
| BC047409.1 | - | 3 | 337 | 5297277 | ||
| BG721781.1 | - | 4 | 471 | |||