Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK3 SIRNA Assays ›
Gene Symbol:
MARK3
Gene Name:
microtubule affinity regulating kinase 3
Gene Aliases:
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location:
Chr.14: 103385364 - 103503831 on Build GRCh38
UniGene ID:
Hs.35828
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS106338
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128918.2
NP_001122390.2
11
1732
NM_001128919.2
NP_001122391.2
11
1732
NM_001128920.2
NP_001122392.2
11
1732
NM_001128921.2
NP_001122393.2
10
1495
NM_002376.6
NP_002367.5
11
1732
XM_005267641.4
XP_005267698.2
11
1111
XM_005267642.4
XP_005267699.2
11
1732
XM_005267643.4
XP_005267700.2
11
1732
XM_006720146.3
XP_006720209.1
11
1106
XM_011536769.2
XP_011535071.1
10
845
XM_017021291.1
XP_016876780.1
11
1732
XM_017021292.1
XP_016876781.1
11
1732
XM_017021293.1
XP_016876782.1
12
1207
XM_017021294.1
XP_016876783.1
10
1481
XM_017021295.1
XP_016876784.1
10
1496
XM_017021296.1
XP_016876785.1
10
1513
XM_017021297.1
XP_016876786.1
10
1489
XM_017021298.1
XP_016876787.1
10
1489
XM_017021299.1
XP_016876788.1
10
1540
XM_017021300.1
XP_016876789.1
9
966
XM_017021301.1
XP_016876790.1
9
1121
XM_017021302.1
XP_016876791.1
9
966
XM_017021303.1
XP_016876792.1
9
966
XM_017021304.1
XP_016876793.1
9
966
XM_017021305.1
XP_016876794.1
9
966
XM_017021306.1
XP_016876795.1
10
1661
XM_017021307.1
XP_016876796.1
9
1612
XM_017021308.1
XP_016876797.1
10
1683
XM_017021309.1
XP_016876798.1
9
944
XM_017021310.1
XP_016876799.1
9
944
XM_017021311.1
XP_016876800.1
8
895
GenBank mRNA
AF170723.1
-
2
169
AF387637.1
-
11
1084
AF465413.1
-
11
1576
AK296643.1
-
4
369
AK307448.1
-
11
1111
BC024773.1
-
11
1555
5138958
BX161395.1
-
10
1425
BX248250.1
-
2
102
M80359.1
-
11
1237
U64205.1
-
11
1441
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
establishment of cell polarity
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-nbkcc:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0