Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107661
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
36
6108
NM_006197.3
NP_006188.3
36
6132
XM_006716336.3
XP_006716399.1
39
6583
XM_006716340.3
XP_006716403.1
39
6571
XM_006716349.3
XP_006716412.1
35
6043
XM_011544528.2
XP_011542830.1
39
6604
XM_011544529.2
XP_011542831.1
41
6910
XM_011544530.2
XP_011542832.1
40
6726
XM_011544532.2
XP_011542834.1
39
6601
XM_011544533.2
XP_011542835.1
39
6592
XM_011544534.2
XP_011542836.1
39
6580
XM_011544535.2
XP_011542837.1
39
6568
XM_011544540.2
XP_011542842.1
39
6532
XM_017013472.1
XP_016868961.1
40
6720
XM_017013473.1
XP_016868962.1
40
6716
XM_017013474.1
XP_016868963.1
37
6334
XM_017013475.1
XP_016868964.1
39
6458
XM_017013476.1
XP_016868965.1
36
6042
XM_017013477.1
XP_016868966.1
39
6623
XM_017013478.1
XP_016868967.1
39
6601
XM_017013479.1
XP_016868968.1
39
6602
XM_017013480.1
XP_016868969.1
37
6274
XM_017013481.1
XP_016868970.1
39
6577
XM_017013482.1
XP_016868971.1
36
6238
XM_017013483.1
XP_016868972.1
38
6507
XM_017013484.1
XP_016868973.1
35
5933
XM_017013485.1
XP_016868974.1
38
6487
XM_017013486.1
XP_016868975.1
36
6169
XM_017013487.1
XP_016868976.1
36
6160
XM_017013488.1
XP_016868977.1
36
6157
XM_017013489.1
XP_016868978.1
38
6460
XM_017013490.1
XP_016868979.1
38
6436
XM_017013491.1
XP_016868980.1
35
6055
XM_017013492.1
XP_016868981.1
35
6052
XM_017013493.1
XP_016868982.1
37
6355
XM_017013494.1
XP_016868983.1
35
6040
XM_017013495.1
XP_016868984.1
37
6180
XM_017013496.1
XP_016868985.1
34
5763
XM_017013497.1
XP_016868986.1
35
6028
XM_017013498.1
XP_016868987.1
35
6019
XM_017013499.1
XP_016868988.1
37
6159
XM_017013500.1
XP_016868989.1
35
6016
XM_017013501.1
XP_016868990.1
37
6318
XM_017013502.1
XP_016868991.1
34
5739
GenBank mRNA
AF068293.1
-
5
633
AK091406.1
-
13
1817
AK295856.1
-
14
2118
AK301727.1
-
13
1624
AK302378.1
-
15
1929
AW080255.1
-
4
462
AX785390.1
-
34
5710
BC140946.1
-
36
5937
9056758
BI966701.1
-
4
456
L27841.1
-
36
6119
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bwhjp:80/100.66.76.134:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0