Search
Search

SEC14L1| Gene Symbol: SEC14L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039573.2 | NP_001034662.2 | 6 | 772 | ||
| NM_001143998.1 | NP_001137470.1 | 6 | 772 | |||
| NM_001143999.1 | NP_001137471.1 | 6 | 768 | |||
| NM_001144001.1 | NP_001137473.1 | 4 | 560 | |||
| NM_001204408.1 | NP_001191337.1 | 8 | 1117 | |||
| NM_001204410.1 | NP_001191339.1 | 7 | 810 | |||
| NM_003003.3 | NP_002994.3 | 6 | 719 | |||
| GenBank mRNA | AB208811.1 | - | 6 | 751 | ||
| AK290913.1 | - | 6 | 665 | |||
| AK293206.1 | - | 4 | 560 | |||
| BC136523.1 | - | 6 | 607 | 9020513 | ||
| BC136525.1 | - | 6 | 607 | 9020515 | ||
| BC142979.1 | - | 6 | 663 | 8860414 | ||
| BM802278.1 | - | 293 | ||||
| BX537448.1 | - | 6 | 673 | |||
| D67029.1 | - | 6 | 755 | |||
| FM995495.1 | - | 8 | 1011 | |||
| FM995496.1 | - | 7 | 808 | |||
| FM995497.1 | - | 7 | 907 | |||