Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110547
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
14
1960
NM_001146283.1
NP_001139755.1
13
1959
NM_001146284.1
NP_001139756.1
13
1857
NM_001146285.1
NP_001139757.1
13
1891
NM_001146286.1
NP_001139758.1
12
1818
NM_001198525.1
NP_001185454.1
15
2030
NM_001198526.1
NP_001185455.1
13
1891
NM_001198527.1
NP_001185456.1
15
1955
NM_001198528.1
NP_001185457.1
15
1967
NM_001198529.1
NP_001185458.1
13
1869
NM_001198530.1
NP_001185459.1
11
1716
NM_001198531.1
NP_001185460.1
13
1887
NM_030756.4
NP_110383.2
14
1942
XM_005270071.1
XP_005270128.1
16
2201
XM_005270075.1
XP_005270132.1
16
2186
XM_005270077.1
XP_005270134.1
15
2150
XM_005270078.1
XP_005270135.1
15
2150
XM_005270079.1
XP_005270136.1
15
2135
XM_005270080.1
XP_005270137.1
15
2132
XM_005270084.1
XP_005270141.1
15
2042
XM_005270085.1
XP_005270142.1
14
2006
XM_005270086.1
XP_005270143.1
14
1988
XM_005270088.1
XP_005270145.1
14
1961
XM_005270089.1
XP_005270146.1
13
1937
XM_005270091.2
XP_005270148.1
17
2274
XM_005270093.2
XP_005270150.1
15
2128
XM_005270094.2
XP_005270151.1
15
2125
XM_005270095.1
XP_005270152.1
14
2077
XM_005270096.2
XP_005270153.1
14
1969
XM_005270100.1
XP_005270157.1
13
1933
XM_005270101.2
XP_005270158.1
13
1915
XM_005270102.1
XP_005270159.1
13
1906
XM_005270103.1
XP_005270160.1
12
1864
XM_005270104.1
XP_005270161.1
12
1837
XM_011540109.1
XP_011538411.1
16
2201
XM_011540110.1
XP_011538412.1
15
2105
XM_011540111.1
XP_011538413.1
14
1973
XM_011540116.1
XP_011538418.1
16
2223
XM_017016584.1
XP_016872073.1
16
2198
XM_017016585.1
XP_016872074.1
16
2189
XM_017016586.1
XP_016872075.1
16
2186
XM_017016587.1
XP_016872076.1
16
2174
XM_017016588.1
XP_016872077.1
15
2132
XM_017016589.1
XP_016872078.1
14
2078
XM_017016590.1
XP_016872079.1
15
2057
XM_017016591.1
XP_016872080.1
15
2045
XM_017016592.1
XP_016872081.1
15
2042
XM_017016593.1
XP_016872082.1
14
1991
XM_017016594.1
XP_016872083.1
14
1988
XM_017016595.1
XP_016872084.1
14
2062
XM_017016596.1
XP_016872085.1
14
1957
GenBank mRNA
AB440195.1
-
13
1380
AB451266.1
-
14
1435
AK074705.1
-
4
333
AK225809.1
-
13
1895
AK299295.1
-
12
1348
AK301483.1
-
11
1158
AK315243.1
-
13
1625
AK316111.1
-
12
1045
BC032656.1
-
13
1446
5533185
CR536574.1
-
14
1435
FJ010164.1
-
13
1330
FJ010165.1
-
14
1462
FJ010166.1
-
15
1616
FJ010167.1
-
13
1477
FJ010170.1
-
13
1455
FJ010171.1
-
11
1246
FJ010172.1
-
12
1408
FJ010173.1
-
13
1330
FJ010174.1
-
14
1839
HM352838.1
-
13
1377
HM352839.1
-
13
1399
HM352841.1
-
13
1395
HM352842.1
-
12
1311
HM352843.1
-
12
1299
HM352844.1
-
14
1431
HM352845.1
-
15
1564
HM352846.1
-
13
1380
HM352847.1
-
13
1384
HM352848.1
-
14
1435
HM352849.1
-
12
1326
HM352850.1
-
15
1504
HM352851.1
-
14
1468
Y11306.2
-
14
1742
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191052.
This siRNA also targets the following transcripts in Mouse: NM_001142919, NM_001142922, NM_009333, NM_001142918, NM_001142923, and others.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0