Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNNT3 SIRNA Assays ›
          Gene Symbol:
          TNNT3
          Gene Name:
          troponin T3, fast skeletal type
          Gene Aliases:
          TNTF
          Chromosome Location:
          Chr.11: 1919551 - 1938706 on Build GRCh38
          UniGene ID:
          Hs.73454
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110886
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNNT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042780.2 NP_001036245.1 13, 14 858
          NM_001042781.2 NP_001036246.1 14, 15 876
          NM_001042782.2 NP_001036247.1 13, 14 858
          NM_001297646.1 NP_001284575.1 14, 15 1116
          NM_006757.3 NP_006748.1 14, 15 882
          XM_006718288.3 XP_006718351.1 9, 10 909
          XM_006718290.3 XP_006718353.1 16, 17 821
          XM_006718293.2 XP_006718356.1 15, 16 898
          XM_006718294.3 XP_006718357.1 15, 16 898
          XM_006718296.3 XP_006718359.1 14, 15 785
          XM_006718299.2 XP_006718362.1 13, 14 721
          XM_006718300.3 XP_006718363.1 12, 13 749
          XM_006718302.3 XP_006718365.1 11, 12 731
          XM_011520343.2 XP_011518645.1 16, 17 952
          XM_017018205.1 XP_016873694.1 14, 15 876
          XM_017018206.1 XP_016873695.1 16, 17 744
          XM_017018207.1 XP_016873696.1 15, 16 732
          XM_017018208.1 XP_016873697.1 14, 15 699
          XM_017018209.1 XP_016873698.1 14, 15 708
          XM_017018210.1 XP_016873699.1 13, 14 673
          XM_017018211.1 XP_016873700.1 13, 14 692
          XM_017018212.1 XP_016873701.1 12, 13 664
          XM_017018213.1 XP_016873702.1 11, 12 649
          XM_017018214.1 XP_016873703.1 11, 12 648
          XM_017018215.1 XP_016873704.1 11, 12 640
          XM_017018216.1 XP_016873705.1 15, 16 803
          XM_017018217.1 XP_016873706.1 14, 15 782
          XM_017018218.1 XP_016873707.1 13, 14 770
          XM_017018219.1 XP_016873708.1 11, 12 731
          GenBank mRNA AI161366.1 - 2, 3 320
          AJ571879.1 - 1, 2 33
          AK056968.1 - 13, 14 937
          BC050446.1 - 13, 14 714
          BC117327.1 - 12, 13 742 40125878
          BC143537.1 - 12, 13 742 9052047
          BC171727.1 - 13, 14 760 9052046
          BC171728.1 - 9, 10 695
          BT019997.1 - 13, 14 664
          BX640689.1 - 14, 15 740
          CR541927.1 - 13, 14 664
          DQ778624.1 - 9, 10 646
          M21984.1 - 13, 14 682

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          regulation of striated muscle contraction
          muscle filament sliding
          regulation of ATPase activity
          actin binding
          tropomyosin binding
          troponin C binding
          calcium-dependent ATPase activity
          troponin I binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0