Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNFSF11 SIRNA Assays ›
          Gene Symbol:
          TNFSF11
          Gene Name:
          tumor necrosis factor superfamily member 11
          Gene Aliases:
          CD254, ODF, OPGL, OPTB2, RANKL, TNLG6B, TRANCE, hRANKL2, sOdf
          Chromosome Location:
          Chr.13: 42562736 - 42608013 on Build GRCh38
          UniGene ID:
          Hs.333791
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS112578
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNFSF11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003701.3 NP_003692.1 5 906
          NM_033012.3 NP_143026.1 7 1067
          XM_011535280.2 XP_011533582.1 5 611
          XM_017020802.1 XP_016876291.1 5 713
          XM_017020803.1 XP_016876292.1 5 560
          GenBank mRNA AB037599.1 - 5 632
          AB061227.1 - 6 714
          AB064268.1 - 6 621
          AB064269.1 - 5 837
          AB064270.1 - 6 775
          AF013171.1 - 4 541
          AF019047.1 - 5 885
          AF053712.1 - 5 941
          BC040889.1 - 7 1067
          BC074823.2 - 5 787 30915538
          BC074890.2 - 5 787 30915245
          BC117286.1 - 5 789 40125837
          BC117288.1 - 5 789 40125839

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoclast proliferation
          monocyte chemotaxis
          immune response
          activation of JUN kinase activity
          organ morphogenesis
          cytokine-mediated signaling pathway
          osteoclast differentiation
          tumor necrosis factor-mediated signaling pathway
          mammary gland epithelial cell proliferation
          positive regulation of homotypic cell-cell adhesion
          paracrine signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of MAP kinase activity
          bone resorption
          positive regulation of osteoclast differentiation
          positive regulation of bone resorption
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of T cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein homooligomerization
          positive regulation of corticotropin-releasing hormone secretion
          positive regulation of protein kinase B signaling
          calcium ion homeostasis
          mammary gland alveolus development
          ERK1 and ERK2 cascade
          positive regulation of fever generation by positive regulation of prostaglandin secretion
          TNFSF11-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade via TNFSF11-mediated signaling
          positive regulation of intracellular signal transduction
          positive regulation of osteoclast development
          cytokine activity
          tumor necrosis factor receptor binding
          tumor necrosis factor receptor superfamily binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0