Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113053
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
11
1746
NM_001145848.1
NP_001139320.1
11
1462
NM_001145849.1
NP_001139321.1
11
1466
NM_001145850.1
NP_001139322.1
11
1466
NM_001145851.1
NP_001139323.1
10
1439
NM_001145852.1
NP_001139324.1
10
1439
NM_006017.2
NP_006008.1
11
1466
XM_005248195.4
XP_005248252.1
11
1562
XM_005248196.4
XP_005248253.1
11
1562
XM_006713974.3
XP_006714037.1
10
1067
XM_011513890.1
XP_011512192.1
12
1548
XM_011513892.2
XP_011512194.1
12
1699
XM_011513893.2
XP_011512195.1
12
1773
XM_011513894.2
XP_011512196.1
12
1858
XM_011513895.2
XP_011512197.1
13
1974
XM_011513896.2
XP_011512198.1
12
1603
XM_011513897.2
XP_011512199.1
12
1790
XM_011513898.2
XP_011512200.2
12
1548
XM_011513899.2
XP_011512201.2
11
1562
XM_011513900.2
XP_011512202.1
12
1548
XM_011513902.2
XP_011512204.1
12
1548
XM_011513903.2
XP_011512205.1
11
1097
XM_017008799.1
XP_016864288.1
11
1592
XM_017008800.1
XP_016864289.1
12
1548
XM_017008801.1
XP_016864290.1
12
1548
XM_017008802.1
XP_016864291.1
12
1640
XM_017008803.1
XP_016864292.1
11
1537
XM_017008804.1
XP_016864293.1
11
1607
XM_017008805.1
XP_016864294.1
11
1480
GenBank mRNA
AF027208.1
-
11
1291
AF117225.1
-
11
1507
AF507034.1
-
10
1264
AK027420.1
-
11
1485
AK027422.1
-
11
1507
AK075411.1
-
11
1462
AK226033.1
-
11
1485
AK310415.1
-
11
1312
AK314529.1
-
11
1507
AK315889.1
-
8
782
AY449689.1
-
10
1227
AY449690.1
-
10
1227
AY449691.1
-
10
1227
AY449692.1
-
11
1254
AY449693.1
-
11
1254
BC012089.1
-
11
1462
4644690
HQ628626.1
-
10
1227
HQ628627.1
-
12
1483
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0