Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIAS2 SIRNA Assays ›
Gene Symbol:
PIAS2
Gene Name:
protein inhibitor of activated STAT 2
Gene Aliases:
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4
Chromosome Location:
Chr.18: 46803224 - 46920167 on Build GRCh38
UniGene ID:
Hs.57769
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS113396
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIAS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324046.1
NP_001310975.1
11
1507
NM_001324047.1
NP_001310976.1
10
1418
NM_001324048.1
NP_001310977.1
11
1507
NM_001324049.1
NP_001310978.1
12
1630
NM_001324051.1
NP_001310980.1
10
1418
NM_001324052.1
NP_001310981.1
12
1608
NM_001324053.1
NP_001310982.1
12
1767
NM_001324054.1
NP_001310983.1
10
1398
NM_001324055.1
NP_001310984.1
11
1526
NM_001324057.1
NP_001310986.1
12
1608
NM_001324058.1
NP_001310987.1
9
1309
NM_001324059.1
NP_001310988.1
11
1507
NM_001324060.1
NP_001310989.1
9
1207
NM_004671.4
NP_004662.2
9
1309
NM_173206.3
NP_775298.1
9
1309
NR_136684.1
8
1224
NR_136685.1
8
834
XM_005258377.3
XP_005258434.1
9
1207
XM_005258379.3
XP_005258436.1
9
1324
XM_006722571.3
XP_006722634.1
10
1292
XM_006722572.3
XP_006722635.1
10
1292
XM_006722573.2
XP_006722636.1
10
1292
XM_011526258.2
XP_011524560.1
9
1372
XM_011526259.2
XP_011524561.1
10
1223
XM_017026068.1
XP_016881557.1
12
1608
XM_017026069.1
XP_016881558.1
10
1341
XM_017026070.1
XP_016881559.1
12
1608
XM_017026071.1
XP_016881560.1
10
1223
XM_017026072.1
XP_016881561.1
9
1162
GenBank mRNA
AF077953.1
-
9
1122
AF077954.1
-
9
1122
AF361054.1
-
9
1200
AK294801.1
-
8
807
AK308143.1
-
8
1146
BC015190.1
-
9
1280
4080297
BC111060.1
-
9
1251
5745101
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
protein sumoylation
androgen receptor signaling pathway
regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of androgen receptor signaling pathway
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
zinc ion binding
ligase activity
SUMO transferase activity
ubiquitin protein ligase binding
androgen receptor binding
SUMO ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0