Search
Search

IL27RA| Gene Symbol: IL27RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004843.3 | NP_004834.1 | 4 | 599 | ||
| GenBank mRNA | AF053004.1 | - | 4 | 604 | ||
| AF106912.1 | - | 4 | 591 | |||
| AF265242.1 | - | 4 | 577 | |||
| AY358876.1 | - | 4 | 577 | |||
| BC028003.1 | - | 4 | 522 | 5212139 | ||