Search
Search

TOB1| Gene Symbol: TOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243877.1 | NP_001230806.1 | 3 | 1014 | ||
| NM_001243885.1 | NP_001230814.1 | 2 | 825 | |||
| NM_005749.3 | NP_005740.1 | 2 | 1028 | |||
| GenBank mRNA | AK313904.1 | - | 3 | 1015 | ||
| AY524047.1 | - | 1 | 595 | |||
| BC031406.1 | - | 1 | 592 | 5194914 | ||
| BC070493.1 | - | 2 | 1016 | 6470511 | ||
| BC098415.1 | - | 2 | 1020 | 6480318 | ||
| BT019380.1 | - | 1 | 585 | |||
| BT019381.1 | - | 1 | 585 | |||
| CR536487.1 | - | 1 | 585 | |||
| CR541686.1 | - | 1 | 585 | |||
| D38305.1 | - | 1 | 628 | |||