Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POMT1 SIRNA Assays ›
          Gene Symbol:
          POMT1
          Gene Name:
          protein O-mannosyltransferase 1
          Gene Aliases:
          LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
          Chromosome Location:
          Chr.9: 131502895 - 131523806 on Build GRCh38
          UniGene ID:
          Hs.522449
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS116296
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POMT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077365.1 NP_001070833.1 17, 18 1909
          NM_001077366.1 NP_001070834.1 16, 17 1757
          NM_001136113.1 NP_001129585.1 17, 18 2149
          NM_001136114.1 NP_001129586.1 15, 16 1650
          NM_007171.3 NP_009102.3 17, 18 1975
          XM_005272156.1 XP_005272213.1 17, 18 2101
          XM_005272158.1 XP_005272215.1 16, 17 1830
          XM_005272159.1 XP_005272216.1 15, 16 1720
          XM_005272162.2 XP_005272219.1 16, 17 2151
          XM_006716932.1 XP_006716995.1 16, 17 1753
          XM_011518140.1 XP_011516442.1 16, 17 1828
          XM_011518141.1 XP_011516443.1 16, 17 1762
          XM_011518142.1 XP_011516444.1 15, 16 1630
          XM_011518143.1 XP_011516445.1 15, 16 1660
          XM_011518145.1 XP_011516447.1 14, 15 1496
          XM_017014203.1 XP_016869692.1 17, 18 2423
          XM_017014204.1 XP_016869693.1 16, 17 1697
          XM_017014205.1 XP_016869694.1 17, 18 1902
          GenBank mRNA AB209430.1 - 11, 12 3605
          AF095136.1 - 17, 18 1952
          AK000475.1 - 6, 7 564
          AK074874.1 - 17, 18 1867
          AK074888.1 - 16, 17 1741
          AK094558.1 - 7, 8 2257
          AK293529.1 - 15, 16 1562
          AK295561.1 - 15, 16 1654
          AK299646.1 - 14, 15 1461
          AK300393.1 - 16, 17 1777
          AK315218.1 - 16, 17 2112
          BC022877.2 - 17, 18 1803 4826806
          BC065268.1 - 17, 18 2094 6155142

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cell wall mannoprotein biosynthetic process
          carbohydrate metabolic process
          protein O-linked glycosylation
          multicellular organism development
          extracellular matrix organization
          protein O-linked mannosylation
          chain elongation of O-linked mannose residue
          regulation of endoplasmic reticulum unfolded protein response
          mannosyltransferase activity
          dolichyl-phosphate-mannose-protein mannosyltransferase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0