Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SULF1 SIRNA Assays ›
          Gene Symbol:
          SULF1
          Gene Name:
          sulfatase 1
          Gene Aliases:
          SULF-1
          Chromosome Location:
          Chr.8: 69466624 - 69660912 on Build GRCh38
          UniGene ID:
          Hs.409602
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118380
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SULF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128204.1 NP_001121676.1 21, 22 3135
          NM_001128205.1 NP_001121677.1 22, 23 3303
          NM_001128206.1 NP_001121678.1 21, 22 3141
          NM_015170.2 NP_055985.2 22, 23 3297
          XM_006716442.1 XP_006716505.1 15, 16 1930
          GenBank mRNA AB029000.1 - 17, 18 2421
          AF545571.1 - 22, 23 3286
          AK055467.1 - 3, 4 1194
          AK074873.1 - 13, 14 1657
          AK123744.1 - 5, 6 458
          AK307951.1 - 21, 22 2915
          AY101175.1 - 21, 22 3088
          BC012997.2 - 2, 3 87
          BX571746.1 - 22, 23 3248

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          negative regulation of endothelial cell proliferation
          chondrocyte development
          glomerular filtration
          apoptotic process
          positive regulation of vascular endothelial growth factor production
          esophagus smooth muscle contraction
          negative regulation of angiogenesis
          positive regulation of Wnt signaling pathway
          heparan sulfate proteoglycan metabolic process
          negative regulation of cell migration
          positive regulation of BMP signaling pathway
          glomerular basement membrane development
          glial cell-derived neurotrophic factor receptor signaling pathway
          regulation of fibroblast growth factor receptor signaling pathway
          negative regulation of fibroblast growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          embryonic skeletal system development
          cartilage development
          bone development
          innervation
          negative regulation of prostatic bud formation
          arylsulfatase activity
          calcium ion binding
          N-acetylglucosamine-6-sulfatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0