Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMUG1 SIRNA Assays ›
Gene Symbol:
SMUG1
Gene Name:
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases:
FDG, HMUDG, UNG3
Chromosome Location:
Chr.12: 54171815 - 54188994 on Build GRCh38
UniGene ID:
Hs.632721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS118999
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMUG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243787.1
NP_001230716.1
4
557
NM_001243788.1
NP_001230717.1
3
473
NM_014311.2
NP_055126.1
4
560
XM_006719319.3
XP_006719382.1
5
659
XM_006719320.3
XP_006719383.1
4
766
XM_006719321.3
XP_006719384.1
3
511
XM_011538109.2
XP_011536411.1
5
830
XM_011538110.2
XP_011536412.1
5
833
XM_011538111.2
XP_011536413.1
4
734
XM_011538112.2
XP_011536414.1
4
736
XM_011538113.2
XP_011536415.1
4
599
XM_011538114.2
XP_011536416.1
4
755
XM_011538115.2
XP_011536417.1
4
629
XM_011538116.2
XP_011536418.1
4
569
XM_011538117.2
XP_011536419.1
4
545
XM_011538118.2
XP_011536420.1
4
590
XM_011538119.2
XP_011536421.1
4
532
XM_011538120.2
XP_011536422.1
3
510
XM_011538121.2
XP_011536423.1
5
834
XM_017019113.1
XP_016874602.1
5
824
XM_017019114.1
XP_016874603.1
3
907
XM_017019115.1
XP_016874604.1
4
632
XM_017019116.1
XP_016874605.1
6
1086
GenBank mRNA
AF125182.1
-
2
442
AK001235.1
-
4
539
AK001789.1
-
3
1564
AK222473.1
-
4
536
AK290274.1
-
3
465
BG706635.1
-
3
476
BG764502.1
-
5
643
CD245413.1
-
2
437
DA430734.1
-
3
465
KU178579.1
-
2
410
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
base-excision repair
depyrimidination
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
DNA binding
uracil DNA N-glycosylase activity
protein binding
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0