Search
Search

REXO2| Gene Symbol: REXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015523.3 | NP_056338.2 | 2 | 342 | ||
| GenBank mRNA | AF151872.1 | - | 2 | 293 | ||
| AK223200.1 | - | 2 | 257 | |||
| AK312042.1 | - | 2 | 271 | |||
| AL110239.1 | - | 2 | 276 | |||
| BC003502.1 | - | 2 | 244 | |||
| BC105024.1 | - | 2 | 263 | 8144027 | ||
| BC105026.1 | - | 2 | 263 | 8144029 | ||
| BC107887.1 | - | 252 | 6663083 | |||
| BC143701.1 | - | 2 | 266 | 9052215 | ||
| CR533500.1 | - | 2 | 199 | |||
| DB471224.1 | - | 2 | 343 | |||