This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
CRBN
- Gene Name:
-
cereblon
- Gene Aliases:
-
MRT2, MRT2A
- Chromosome Location:
-
Chr.3: 3148490 - 3179717 on Build GRCh38
- UniGene ID:
-
Hs.18925
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
CRBN |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001173482.1 |
NP_001166953.1 |
2, 3
|
189
|
|
| NM_016302.3 |
NP_057386.2 |
2, 3
|
192
|
|
| XM_011533791.2 |
XP_011532093.1 |
2, 3
|
179
|
|
| XM_011533793.2 |
XP_011532095.1 |
2, 3
|
179
|
|
| GenBank mRNA |
AF117230.1 |
- |
2, 3
|
156
|
|
| AK027507.1 |
- |
2, 3
|
170
|
|
| AK058191.1 |
- |
2, 3
|
166
|
|
| AK312577.1 |
- |
2, 3
|
170
|
|
| AL711072.1 |
- |
2, 3
|
183
|
|
| BC017419.1 |
- |
2, 3
|
180
|
4672890 |
| BC067811.1 |
- |
2, 3
|
173
|
5264951 |
| DB456585.1 |
- |
2, 3
|
192
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System