Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDH23 SIRNA Assays ›
Gene Symbol:
CDH23
Gene Name:
cadherin-related 23
Gene Aliases:
CDHR23, USH1D
Chromosome Location:
Chr.10: 71396934 - 71815947 on Build GRCh38
UniGene ID:
Hs.656032
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS127316
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDH23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171930.1
NP_001165401.1
11
1392
NM_001171931.1
NP_001165402.1
11
1392
NM_001171932.1
NP_001165403.1
11
1392
NM_022124.5
NP_071407.4
11
1392
NM_052836.3
NP_443068.1
11
1392
XM_006717940.3
XP_006718003.1
11
1386
XM_006717942.3
XP_006718005.1
11
1386
XM_011540039.2
XP_011538341.1
11
1386
XM_011540042.2
XP_011538344.1
11
1386
XM_011540043.2
XP_011538345.1
11
1386
XM_011540044.2
XP_011538346.1
11
1027
XM_011540045.2
XP_011538347.1
11
1386
XM_011540046.2
XP_011538348.1
6
821
XM_011540048.2
XP_011538350.1
11
1386
XM_011540049.2
XP_011538351.1
11
1386
XM_011540051.2
XP_011538353.1
11
1386
XM_017016499.1
XP_016871988.1
11
1392
XM_017016500.1
XP_016871989.1
11
1386
GenBank mRNA
AF312024.1
-
10
1002
AK074160.1
-
4
374
AY010111.2
-
10
1002
AY358617.1
-
11
1042
BC011570.1
-
11
1397
BC032581.1
-
11
1398
BC065284.1
-
11
1369
BC108254.1
-
9
1160
BC136976.1
-
11
1262
BC136977.1
-
11
1261
BC139903.1
-
11
1384
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
homophilic cell adhesion via plasma membrane adhesion molecules
visual perception
sensory perception of sound
locomotory behavior
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
photoreceptor cell maintenance
response to stimulus
sensory perception of light stimulus
equilibrioception
regulation of cytosolic calcium ion concentration
inner ear receptor stereocilium organization
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0