Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MPPE1 SIRNA Assays ›
          Gene Symbol:
          MPPE1
          Gene Name:
          metallophosphoesterase 1
          Gene Aliases:
          PGAP5
          Chromosome Location:
          Chr.18: 11883471 - 11908797 on Build GRCh38
          UniGene ID:
          Hs.712666
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS128021
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MPPE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242904.1 NP_001229833.1 4 1098
          NM_001319154.1 NP_001306083.1 5 1205
          NM_023075.5 NP_075563.3 5 1205
          NR_040241.1 5 1205
          NR_040242.3 6 1073
          NR_040243.1 5 1205
          XM_006722340.3 XP_006722403.1 5 758
          XM_006722341.3 XP_006722404.1 7 1380
          XM_006722342.3 XP_006722405.1 6 1164
          XM_006722343.3 XP_006722406.1 6 1285
          XM_006722345.3 XP_006722408.1 4 1098
          XM_006722346.2 XP_006722409.1 4 1098
          XM_006722347.3 XP_006722410.1 5 758
          XM_006722348.2 XP_006722411.1 5 758
          XM_011525731.2 XP_011524033.1 8 1788
          XM_011525732.1 XP_011524034.1 5 758
          XM_011525734.1 XP_011524036.1 5 758
          XM_017025920.1 XP_016881409.1 7 1693
          XM_017025921.1 XP_016881410.1 4 600
          XM_017025922.1 XP_016881411.1 7 1380
          XM_017025923.1 XP_016881412.1 8 1788
          XM_017025924.1 XP_016881413.1 6 1164
          XM_017025925.1 XP_016881414.1 6 1285
          XM_017025926.1 XP_016881415.1 7 1693
          XM_017025927.1 XP_016881416.1 6 1164
          XM_017025928.1 XP_016881417.1 6 1285
          XM_017025929.1 XP_016881418.1 4 1098
          XM_017025930.1 XP_016881419.1 4 1098
          XM_017025931.1 XP_016881420.1 5 758
          GenBank mRNA AF289560.1 - 5 726
          AF363484.1 - 5 1207
          AK023052.1 - 3 422
          AK054639.1 - 6 571
          AK223450.1 - 4 578
          AK309948.1 - 5 723
          AL833636.1 - 5 757
          AX775853.1 - 5 755
          BC002877.2 - 4 585 3942160
          BC073994.1 - 5 717 6180123

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          ER to Golgi vesicle-mediated transport
          protein binding
          phosphoric diester hydrolase activity
          manganese ion binding
          GPI anchor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-chn84:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0