Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 7, 8 | 1128 | ||
NM_024046.4 | NP_076951.2 | 7, 8 | 1128 | |||
GenBank mRNA | AK074856.1 | - | 7, 8 | 842 | ||
AK075294.1 | - | 7, 8 | 823 | |||
BC000497.2 | - | 7, 8 | 797 | 2820734 | ||
BC001921.1 | - | 7, 8 | 797 | 2820734 | ||
BC005828.2 | - | 7, 8 | 797 | 2820734 | ||
BC017363.2 | - | 7, 8 | 799 | 3940018 | ||
BC019256.1 | - | 7, 8 | 792 | 3502894 |