Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS128710
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
5
471
NM_001164629.3
NP_001158101.1
5
456
NM_001284233.1
NP_001271162.1
3
280
NM_001284234.1
NP_001271163.1
3
171
NM_001284235.1
NP_001271164.1
5
438
NM_001284238.1
NP_001271167.1
7
670
NM_024659.4
NP_078935.2
5
438
XM_005263774.2
XP_005263831.1
5
408
XM_005263782.2
XP_005263839.1
5
424
XM_005263784.2
XP_005263841.1
3
283
XM_011511843.2
XP_011510145.1
5
457
XM_011511855.2
XP_011510157.1
3
401
XM_011511856.2
XP_011510158.1
3
183
XM_017004917.1
XP_016860406.1
9
1079
XM_017004918.1
XP_016860407.1
7
671
XM_017004919.1
XP_016860408.1
5
557
XM_017004920.1
XP_016860409.1
7
3713
XM_017004921.1
XP_016860410.1
6
506
XM_017004922.1
XP_016860411.1
6
683
XM_017004923.1
XP_016860412.1
5
471
XM_017004924.1
XP_016860413.1
7
667
XM_017004925.1
XP_016860414.1
4
329
XM_017004926.1
XP_016860415.1
3
407
XM_017004927.1
XP_016860416.1
5
438
XM_017004928.1
XP_016860417.1
7
671
XM_017004929.1
XP_016860418.1
5
408
XM_017004930.1
XP_016860419.1
5
1059
XM_017004931.1
XP_016860420.1
5
472
XM_017004932.1
XP_016860421.1
5
442
XM_017004933.1
XP_016860422.1
5
424
XM_017004934.1
XP_016860423.1
6
3631
XM_017004935.1
XP_016860424.1
7
1501
XM_017004936.1
XP_016860425.1
6
678
XM_017004937.1
XP_016860426.1
3
412
XM_017004938.1
XP_016860427.1
3
410
XM_017004939.1
XP_016860428.1
5
460
XM_017004940.1
XP_016860429.1
3
408
XM_017004941.1
XP_016860430.1
3
407
XM_017004942.1
XP_016860431.1
5
454
XM_017004943.1
XP_016860432.1
5
1120
XM_017004944.1
XP_016860433.1
5
453
XM_017004945.1
XP_016860434.1
7
671
XM_017004946.1
XP_016860435.1
5
408
XM_017004947.1
XP_016860436.1
5
1091
XM_017004948.1
XP_016860437.1
6
3631
XM_017004949.1
XP_016860438.1
3
406
XM_017004950.1
XP_016860439.1
5
453
XM_017004951.1
XP_016860440.1
5
408
XM_017004952.1
XP_016860441.1
5
852
XM_017004953.1
XP_016860442.1
5
424
XM_017004954.1
XP_016860443.1
6
512
XM_017004955.1
XP_016860444.1
8
860
XM_017004956.1
XP_016860445.1
3
408
XM_017004957.1
XP_016860446.1
5
454
GenBank mRNA
AB201269.1
-
2
192
AB597184.1
-
2
192
AK091684.1
-
3
284
AK291357.1
-
5
386
AY281366.1
-
5
471
BC017741.2
-
6
478
4390876
BC061699.1
-
5
378
4608518
BX647436.1
-
3
153
BX647773.1
-
7
609
DB458955.1
-
5
438
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline