Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM62 SIRNA Assays ›
Gene Symbol:
TMEM62
Gene Name:
transmembrane protein 62
Gene Aliases:
-
Chromosome Location:
Chr.15: 43133115 - 43185147 on Build GRCh38
UniGene ID:
Hs.511175
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS129286
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM62
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024956.3
NP_079232.3
8
1179
XM_005254682.4
XP_005254739.1
7
1146
XM_005254683.4
XP_005254740.1
6
1035
XM_005254684.3
XP_005254741.1
5
892
XM_011522071.1
XP_011520373.1
7
663
XM_011522072.2
XP_011520374.1
6
830
XM_011522073.1
XP_011520375.1
4
328
XM_011522074.2
XP_011520376.1
7
974
XM_017022600.1
XP_016878089.1
7
1178
XM_017022601.1
XP_016878090.1
7
931
XM_017022602.1
XP_016878091.1
6
1067
XM_017022603.1
XP_016878092.1
7
663
XM_017022604.1
XP_016878093.1
7
1147
XM_017022605.1
XP_016878094.1
6
1036
XM_017022606.1
XP_016878095.1
6
1004
XM_017022607.1
XP_016878096.1
5
924
XM_017022608.1
XP_016878097.1
5
458
XM_017022609.1
XP_016878098.1
6
1005
XM_017022610.1
XP_016878099.1
6
765
XM_017022611.1
XP_016878100.1
6
820
XM_017022612.1
XP_016878101.1
6
939
XM_017022613.1
XP_016878102.1
5
631
XM_017022614.1
XP_016878103.1
5
589
XM_017022615.1
XP_016878104.1
5
570
XM_017022616.1
XP_016878105.1
5
628
GenBank mRNA
AK027028.1
-
5
484
AK225353.1
-
5
484
AK309679.1
-
7
695
BC009981.2
-
8
1179
4121449
CR457370.1
-
3
276
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA helicase
Gene Ontology Categories:
Process(es)
hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0