Search
Search

CPXM2| Gene Symbol: CPXM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198148.2 | NP_937791.2 | 3, 4 | 836 | ||
| XM_005269528.3 | XP_005269585.1 | 3, 4 | 686 | |||
| XM_011539283.2 | XP_011537585.1 | 3, 4 | 686 | |||
| XM_011539285.2 | XP_011537587.1 | 3, 4 | 468 | |||
| XM_011539286.1 | XP_011537588.1 | 2, 3 | 410 | |||
| XM_017015673.1 | XP_016871162.1 | 4, 5 | 437 | |||
| GenBank mRNA | AK296365.1 | - | 3, 4 | 243 | ||
| AK304023.1 | - | 3, 4 | 610 | |||
| AK304814.1 | - | 3, 4 | 647 | |||
| AL833732.1 | - | 1, 2 | 14 | |||
| AY358565.1 | - | 3, 4 | 836 | |||
| BC027623.1 | - | 4, 5 | 382 | |||
| BC036789.1 | - | 3, 4 | 666 | 5752707 | ||
| BC137492.1 | - | 3, 4 | 831 | 9021492 | ||
| BC146862.1 | - | 3, 4 | 831 | 9056790 | ||