Search
Search

GPC2| Gene Symbol: GPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152742.2 | NP_689955.1 | 5, 6 | 1117 | ||
| GenBank mRNA | AK096281.1 | - | 5, 6 | 1055 | ||
| AL834418.1 | - | 5, 6 | 1003 | |||
| BC027972.1 | - | 5, 6 | 1058 | 5240315 | ||