This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
TMEM95
- Gene Name:
-
transmembrane protein 95
- Gene Aliases:
-
UNQ9390
- Chromosome Location:
-
Chr.17: 7354968 - 7357219 on Build GRCh38
- UniGene ID:
-
Hs.259432
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
TMEM95 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001320435.1 |
NP_001307364.1 |
2, 3
|
285
|
|
| NM_001320436.1 |
NP_001307365.1 |
2, 3
|
285
|
|
| NM_198154.2 |
NP_937797.1 |
2, 3
|
285
|
|
| XM_017024565.1 |
XP_016880054.1 |
2, 3
|
307
|
|
| XM_017024568.1 |
XP_016880057.1 |
2, 3
|
307
|
|
| XM_017024569.1 |
XP_016880058.1 |
2, 3
|
307
|
|
| XM_017024571.1 |
XP_016880060.1 |
2, 3
|
453
|
|
| GenBank mRNA |
AY358218.1 |
- |
2, 3
|
243
|
|
| BC040900.1 |
- |
2, 3
|
285
|
|
| BC107110.2 |
- |
2, 3
|
243
|
40022149 |
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System